Learn More
Abeba™ Insoles 3572
Product Code. 15600258
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
Unit Size:
1 pair
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15600258 36 1 pair
15610258 37 1 pair
15620258 38 1 pair
15630258 39 1 pair
15640258 40 1 pair
15650258 41 1 pair
15660258 42 1 pair
15670258 43 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15600258 Supplier Abeba™ Supplier No. 3572/36
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Massage knobs stimulate circulation. ESD dissipative and antistatic.
  • Microfiber
  • Anthracite
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Stimulating circulation by massage knobs, comfortable orthopaedic footbed
  • Reflexor massage active-footbed
Specifications
Color Anthracite
Material Microfiber
Size 36
Style 3572