missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Inspection Windows For Titrette™

Product Code. 10576265 Shop All Brand Products
Change view
Click to view available options
For Use With Titrette™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10576265 For Use With Titrette™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10576265 Supplier BRAND™ Supplier No. 6783
  • ONLINE EXCLUSIVE PRICE
    15.85 €
    14.24 € / Pack of 1
    Save 1.61 €
    10% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes one clear and one amber colored window. For protection of light-sensitive media, replace the clear inspection window with the amber colored window.

  • Plastic construction
  • For use with Titrette™
  • One clear, one amber colored window included
  • Amber colored window prevents damage to light-sensitive media
  • Supplied in packs of two
TRUSTED_SUSTAINABILITY

Specifications

Color Amber, Clear
For Use With (Equipment) Protection of Light-Sensitive Media
Material Plastic
No. per Pack Set of 2
Type Inspection Window
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.