Learn More
JULABO™ Installation Cooling Coil for PURA™ Water Baths
Click to view available options
For Use With (Equipment):
PURA™ 10 and PURA™ 14
PURA™ 22 and PURA™ 30
Unit Size:
Each
Description
- Accessory for PURA™ Water Baths
- For working near ambient temperature
Specifications
Specifications
| Description | Cooling coil |
| For Use With (Equipment) | PURA™ 10 and PURA™ 14 |
| Weight (Metric) | 0.272 kg (net) |