Learn More
Thermo Scientific Instrument for Enterprise
Product Code. 31906282 Shop All Thermo Scientific Products
Change view
Click to view available options
Operating System:
Windows 11
Unit Size:
Each
Product Code. Operating System Unit Size
31906282 Windows 11 Each
1 options
Product Code. 31906282 Supplier Thermo Scientific Supplier No. 7200.1000
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
The Enterprise Instrument license enables full control and data acquisition of one Thermo Scientific instrument under Chromeleon CDS (any version 7.x).
Includes:
  • Instrument Controller
  • Spectral license for 3D data acquisition of diode-array UV and unit-resolution MS spectra
  • Fraction collection
TRUSTED_SUSTAINABILITY
Specifications
Type Chromatography Software
For Use With (Equipment) LC, IC, and GC Instruments and Accessories
Certifications/Compliance 21 CFR
Product Line Chromeleon 7
Operating System Windows 11
Quantity 1 Ea.