missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Insulin B (22-25)

Product Code. 15463789
Change view
Click to view available options
Quantity:
25mg
100mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463789 25mg 25mg
15473789 100mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463789 Supplier Thermo Scientific Chemicals Supplier No. J61170.MB
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

For research use

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized powder
Quantity 25mg
Storage Requirements -30° to -10°C
Regulatory Status RUO
Protein Length H-Arg-gly-Phe-Phe-OH
Product Type Insulin B (22-25)
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.