missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Insulin-Transferrin-Selenium (ITS-G) Select Supplement (100X)

Product Code. 30282859 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30282859 10 mL 10mL
30283560 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30282859 Supplier Gibco™ Supplier No. A4000046501
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Insulin-Transferrin-Selenium (ITS-G) Select Supplement is an animal-origin-free (AOF) basal medium supplement that is used to reduce the amount of FBS needed to culture cells.

Gibco Insulin-Transferrin-Selenium (ITS-G) Select Supplement is an animal origin-free (AOF) basal medium supplement. The insulin and transferrin in the ITS-G Select formulation are produced in non-mammalian recombinant expression systems.

ITS-G is used as a basal medium supplement to reduce the amount of fetal bovine serum (FBS) needed to culture cells. Originally used with RPMI 1640 and Minimum Essential Medium (MEM), ITS-G is now routinely used with other basal media to support a wide variety of cell types in the presence of 2–4% FBS.

Insulin promotes glucose and amino acid uptake, lipogenesis, intracellular transport, and the synthesis of proteins and nucleic acids. Transferrin is an iron carrier and may also help reduce toxic levels of oxygen radicals and peroxide. Selenium, as sodium selenite, is a co-factor for glutathione peroxidase and other proteins and is used as an antioxidant in media.

The product formulation contains the following components:

  • Insulin (1000.0 mg/L)
  • Transferrin (550.0 mg/L)
  • Sodium selenite (0.67 mg/L)

cGMP manufacturing and quality system

ITS-G Select Supplement is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Endotoxin Level ≤ 1.00 EU/mL
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 260 to 320 mOsm/kg
pH 7.8 to 9.0
Quantity 10 mL
Shipping Condition Room Temperature
Source Animal Origin-free
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.