Learn More
Manutan Internal doors, quick hang PVC curtains, 2400mm x 1500mm
Product Code. 12854389
Encompass_Preferred
Product Code. 12854389 Supplier Manutan Supplier No. A352100
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
  • Drive through flexible curtains for internal doors. PVC strips create a see through flexible curtain. Keeps heat in and reduces cold air loss in chill/freezer rooms for greater temperature control. Provides protection against rain, wind and restricts insect and bird access. Forms a good noise barrier with partial (50%) overlap.
  • Dimensions (H x W): 2400mm x 1500mm