missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Internal doors, quick hang PVC curtains, 3600mm x 4000mm
Description
- Drive through flexible curtains for internal doors. PVC strips create a see through flexible curtain. Keeps heat in and reduces cold air loss in chill/freezer rooms for greater temperature control. Provides protection against rain, wind and restricts insect and bird access. Forms a good noise barrier with partial (50%) overlap.
- Dimensions (H x W): 3600mm x 4000mm
Specifications
Specifications
| Dimensions (L x W) | 3600 x 4000 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?