missing translation for 'onlineSavingsMsg'
Learn More

Inulicin, MedChemExpress

Product Code. 16499168
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499168 10 mg 10mg
16401588 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499168 Supplier Medchem Express Supplier No. HYN0896
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Inulicin (1-O-Acetylbritannilactone) is an active compound that inhibits VEGF-mediated activation of Src and FAK. Inulicin (1-O-Acetylbritannilactone) inhibits LPS-induced PGE2 production and COX-2 expression, and NF-κB activation and translocation.

Inulicin (1-O-Acetylbritannilactone) is an active compound that inhibits VEGF-mediated activation of Src and FAK. Inulicin (1-O-Acetylbritannilactone) inhibits LPS-induced PGE2 production and COX-2 expression, and NF-κB activation and translocation.

Specifications

Chemical Name or Material Inulicin
CAS 33627-41-7
Color White
Purity Grade Notes Research
Molecular Formula C17H24O5
Quantity 10 mg
Synonym 1-O-Acetylbritannilactone
Solubility Information DMSO : 100 mg/mL (324.29 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (162.14 mM; Need ultrasonic)
SMILES O[C@H]1[C@@](C2=C)([H])[C@@](OC2=O)([H])CC(C)=C1[C@@H](C)CCCOC(C)=O
Molecular Weight (g/mol) 308.37
Formula Weight 308.37
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.