missing translation for 'onlineSavingsMsg'
Learn More

Inulin

Product Code. 11478347
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478347 10 g 10g
11488347 50 g 50g
11498347 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478347 Supplier Thermo Scientific Alfa Aesar Supplier No. A18425.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9005-80-5

Mixture of fructose polymers that serves as carbohydrate storage in plantsInulin is used in the treatment of high blood fats, cholesterol and triglycerides. It is also used for weight loss, constipation and also serves as a food additive to improve taste and as a vaccine adjuvant. Further, it is utilized to replace sugar, fat and flour. It is also used to measure kidney function by determining the glomerular filtration rate (GFR). In addition to this, it is used to replace some of the flour in baked goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mixture of fructose polymers that serves as carbohydrate storage in plantsInulin is used in the treatment of high blood fats, cholesterol and triglycerides. It is also used for weight loss, constipation and also serves as a food additive to improve taste and as a vaccine adjuvant. Further, it is utilized to replace sugar, fat and flour. It is also used to measure kidney function by determining the glomerular filtration rate (GFR). In addition to this, it is used to replace some of the flour in baked goods.

Solubility
Soluble in water.

Notes
Yields D-fructose and D-glucose after acid hydrolysis.Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-80-5
MDL Number MFCD00131407

Specifications

Assay Percent Range 0.98
Odor Odorless
Quantity 10 g
Merck Index 14,5004
Solubility Information Soluble in water.
Chemical Name or Material Inulin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.