missing translation for 'onlineSavingsMsg'
Learn More

Inulin, TRC

Product Code. 30137911
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137911 10 g 10g
30137347 25 g 25g
30009697 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137911 Supplier Toronto Research Chemicals Supplier No. TRCI66668010G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-80-5
Molecular Formula (C6 H10 O5)n
Synonym Advax, Advax D, Alant starch, Alantin, Beneo, Beneo GR, Beneo HP, Beneo IPS, Beneo ST, Dahlin, Fibruline, Fibruline Instant, Fibruline LC, Fibruline Long Chain, Fibruline XL, Fibrulose, Fibrulose F 97, FruTEX, Frutafit, Frutafit CLR, Frutafit EXL, Frutafit HD, Frutafit IQ, Frutafit TEX, Frutagave PR 95, Frutagave SP 70, Fuji FF, Fuji SS, Inulin HD, Inulin IQ, Inutec N 10, Inutec N 10GR, Inutec N 25, LV 110, ORAFTI Synergy 1, Oliggo-Fiber F 97, Oliggo-Fiber XL, Orafti HP, Orafti HPX, Orafti Synergy, Prebiofeed, Raftifeed IPS, Raftiline, Raftiline GR, Raftiline HP, Raftiline LS, Raftiline ST, Sinantrin, Synantherin, Synanthrin, Synantrin, Ticaloid LC-CSR 2

Specifications

Quantity 10 g
Chemical Name or Material Inulin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.