missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 50 bp DNA Ladder
GREENER_CHOICE

Product Code. 11518626
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11518626 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 11518626 Supplier Invitrogen™ Supplier No. 10416014
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Includes

50bp DNA Ladder (1μg/μL) in 10mM Tris-HCl (pH 7.5), 1mM EDTA

Invitrogen 50 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 50 bp to 2,500 bp.

Invitrogen 50 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 50 bp to 2,500 bp. 50 bp DNA Ladder consists of 17 individual chromatography-purified DNA fragments and has reference bands at 2500, 800, and 350 bp for easy orientation.

50 bp DNA Ladder is ideal for separation on 2% agarose gels.

Highlights of 50 bp DNA Ladder
Sharp, clear bands—chromatography purified fragments for consistent and reliable results
Convenient—provided with 6X TrackIt Cyan/Orange Loading Buffer for tracking of DNA migration
Precise—an exact amount of DNA in each band

Product use
The double-stranded DNA ladder can be visualized on 2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

This ladder can be radiolabeled with T4 polynucleotide kinase or T4 DNA polymerase.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 100 µL 50 bp DNA Ladder
• 1 mL 6X TrackIt Cyan/Orange Loading Buffer

Store at -20°C.

Concentration 0.5 μg/μL
Gel Type Agarose
Ready to Load No
Sample Loading Volume 1 mL
Size Range 50 to 2500 bp
Volume (Metric) 100 μL
Gel Compatibility Agarose gel
No. of Reactions 100 Applications
Quantity 50 μg
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Type DNA Ladder
Show More Show Less
Can I know the sequences of Invitrogen DNA ladders?

Sequences of Invitrogen DNA and RNA ladders are proprietary.

Are Invitrogen DNA ladders composed of linear or circular/supercoiled DNA?

Invitrogen DNA ladders contain linear dsDNA fragments.

Are Invitrogen DNA ladders composed of single-stranded or double-stranded DNA fragments?

Invitrogen DNA ladders are composed of double-stranded DNA fragments only.

I'm seeing anomalous migration of my DNA ladder. What happened?

This can happen if the marker was heated. Please ensure that the ladders are not heated before use.

What is the difference between the TrackIt DNA Ladders and other DNA ladders?

TrackIt DNA Ladders contain two tracking dyes in the sample buffers, which serve as visual markers for tracking electrophoresis progress through the gel, and also to indicate when maximum resolution is achieved. The tracking dyes should not obscure your visualization of DNA bands in the ladder, as the dyes run outside the limits of most DNA bands in the ladder.

TrackIt Cyan/Orange Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Orange G. We recommend TrackIt Cyan/Orange Loading Buffer for DNA fragments between 10 bp and 1 kb.

The TrackIt Cyan/Yellow Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Tartrazine. We recommend TrackIt Cyan/Yellow Loading Buffer for DNA fragments between 100 bp and 10 kb. The molecular weights are Xylene Cyanol FF, 638.6; Orange G, 452.4; Tartrazine, 534.4.

Note: The TrackIt DNA Ladders are not recommended for use with polyacrylamide gels and are not designed for quantitation.

I am using a quantitative DNA standard and see little difference between the bands when they are ethidium bromide stained. How can this occur and what can I do?

Here are a few suggestions:

(1) Excess volume loaded on gel. The smallest loading volume will result in the greatest difference in band thickness between different masses. Basically overloading the ladders gives saturated bands which do not show any variation in intensity. It is best to load all samples in the same volume.

(2) Sample loaded in larger or smaller volume than standard. Load equal volumes of sample and standard DNA for accurate comparison.

(3) Ethidium bromide (EtBr) staining after electrophoresis. Include ethidium bromide in the gel rather than staining after electrophoresis to increase the difference in band intensity.

(4) EtBr was in the gel, but not the running buffer. This results in the absence of EtBr in the lower half of the gel after electrophoresis and therefore differential fluorescence between bands in the upper half of the gel and the lower half of the gel.

What can cause anomalous migration of my DNA markers?

In general there are very few reasons why a DNA marker would run anomalously. Here are a few suggestions:

(1) For Lambda DNA/Hind III Fragments, the cos site reannealed. Heat Lambda DNA/Hind III Fragments at 65 degrees C for no longer than 10 min before electrophoresis.

(2) Improper electrophoresis conditions were used. Do not allow voltage to exceed ~20 V/cm. Maintain a temperature <30 degrees C during electrophoresis. Check that the electrophoresis buffer used has sufficient buffering capacity.

(3) DNA was denatured. Do not heat standards (except Lambda-derived markers) prior to electrophoresis. Dilute markers in a buffer containing 20 mM NaCl.

I ran my DNA molecular weight ladder, and several bands seem to be missing. Can you offer suggestions on how to rectify this?

There are a few possible reasons for this:

(1) Small DNA bands were electrophoresed off the gel. Electrophorese the gel for less time, at lower voltage, or use a higher percentage gel.

(2) The small bands migrated with the dye front due to differences in ionic strength between gel and buffer. Be sure the buffer in the gel is the same as the electrophoresis buffer.

(3) DNA bands of similar molecular size were not resolved. Increase the electrophoresis time and check the proper percentage gel for resolution. For DNA fragments <1,000 bp, try Agarose 1000 (Cat. No. 16550100) instead of agarose.

(4) DNA was denatured. Do not heat standards (except Lambda-derived markers) prior to electrophoresis. Dilute markers in a buffer containing 20 mM NaCl.

Are there any suggestions to improve the detection of the bands of my DNA molecular weight ladder in an agarose gel?

Faint or absent DNA ladders might be due to:

(1) An insufficient quantity or concentration of DNA loaded on the gel. You can increase the amount of DNA.

(2) Degraded DNA. Avoid nuclease contamination of the DNA standards. Use sterile or autoclaved tips.

(3) DNA was electrophoresed off the gel. Electrophorese the gel for less time, at a lower voltage, or use a higher percentage gel.

(4) An improper UV light source was used for ethidium bromide - stained DNA. Use short-wavelength (254 nm) UV light for greater sensitivity.

(5) The fluorescence bleached after long UV exposure. Stain the gel again with ethidium bromide.

(6) For Biotinylated Lambda DNA/Hind III Fragments or Biotinylated PhiX174/Hinf I Fragments, the biotin was removed by exposure to alkaline conditions. Minimize exposure of biotinylated DNA to NaOH. Note: Alkaline transfers to a membrane do not present a problem when performed at room temperature.

What could be causing smearing of my molecular weight DNA ladder?

(1) Incorrect concentration of DNA ladder gives poor separation and smearing of DNA bands.
(2) Samples containing ≥50 mM NaCl, 100 mM KCl, 10 mM acetate ions, or 10 mM EDTA (i.e. certain restriction enzyme and PCR buffers) will cause loss of resolution on E-Gel agarose gels.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.