missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lambda Hind III dsDNA Markers

Product Code. 10301095
Click to view available options
Quantity:
200 ug
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10301095

Brand: Invitrogen™ AM7720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Generates fragments of the sizes 23,130, 9416, 6557, 4361, 2322, 2027, 564, and 125 bp.

Generates fragments of the sizes 23,130, 9416, 6557, 4361, 2322, 2027, 564, and 125 bp. These DNA molecular mass markers are supplied with a protocol describing easy labeling of the markers with radioisotopes. They can also be stained with ethidium bromide. One tube contains 200 μg.

Order Info

Shipping Conditions: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA Marker
Content And Storage Store at –20°C.
Gel Compatibility Agarose Gels
Shipping Condition Room Temperature
Size Range 125 to 23.1 kb
Concentration 0.5 mg/mL
Quantity 200 ug
Product Line Ambion™
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.