missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ siPORT™ Amine Transfection Agent with manual

Product Code. 10601196
Click to view available options
Quantity:
400 μL
1 mL
Product Type:
Transfection Reagent
Transfection Reagent with Manual
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10601196

Brand: Invitrogen™ AM4502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Supplied in one tube containing 0.4mL.

Animal origin-free polyamine-based transfection agent for use in transfecting cells during subculturing—saving time, without increasing toxicity

  • Successful siRNA experiment requires the appropriate transfection agent for your cells, and one that is optimized for siRNA delivery
  • Efficiency with which different transfection reagents deliver nucleic acids can vary dramatically with the type of nucleic acid being transfected (RNA vs. DNA, single-stranded vs. double-stranded) and the cells being used

Accessory Products:

  • siPORT™ NeoFX™ Transfection Agent (Mfr. Nos. AM4511 and AM4510), a cationic and neutral lipid mixture, is also available
  • For testing which transfection agent is best with your system, the Silencer™ siRNA Transfection II Kit (Mfr. No. AM1631) which contains the two different transfection agents and controls for transfection optimization, is recommended

Cell Culture, RNAi Transfection, Synthetic siRNA Transfection, Transfection

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free
Content And Storage Store at 4°C.
Cell Type Primary Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Synthetic siRNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line Ambion, siPORT
Product Type Transfection Reagent
Quantity 400 μL
Serum Compatible Yes
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.