missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ siPORT™ Amine Transfection Agent with manual
Click to view available options
Quantity:
400 μL
1 mL
Product Type:
Transfection Reagent
Transfection Reagent with Manual
Unit Size:
Each
Beskrivning
Innehåller
Supplied in one tube containing 0.4mL.
- Successful siRNA experiment requires the appropriate transfection agent for your cells, and one that is optimized for siRNA delivery
- Efficiency with which different transfection reagents deliver nucleic acids can vary dramatically with the type of nucleic acid being transfected (RNA vs. DNA, single-stranded vs. double-stranded) and the cells being used
Accessory Products:
- siPORT™ NeoFX™ Transfection Agent (Mfr. Nos. AM4511 and AM4510), a cationic and neutral lipid mixture, is also available
- For testing which transfection agent is best with your system, the Silencer™ siRNA Transfection II Kit (Mfr. No. AM1631) which contains the two different transfection agents and controls for transfection optimization, is recommended
Cell Culture, RNAi Transfection, Synthetic siRNA Transfection, Transfection
Beställningsinfo
Shipping Condition: Wet Ice
Specifikationer
Specifikationer
| Classification | Animal Origin-Free |
| Content And Storage | Store at 4°C. |
| Cell Type | Primary Cells |
| Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
| Sample Type | Synthetic siRNA |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | Transfection |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Product Line | Ambion, siPORT |
| Product Type | Transfection Reagent |
| Visa mer |
For Research Use Only. Not for use in diagnostic procedures.
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering