missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BenchPro™ 2100 Plasmid Purification Card and Reagent Tray Kit

Product Code. 10398429
Change view
Click to view available options
Quantity:
4 Sets/Box
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10398429 4 Sets/Box Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10398429 Supplier Invitrogen™ Supplier No. MC2001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

BenchPro™2100 Plasmid Purification Cards 1 pack of 4, Cell Liners 1 pack of 4, Cell Liner Lids 1 pack of 4, Elution Tubes (2mL) 1 pack of 4, Reagent Trays (prefilled and foil-sealed with reagents) 4 trays

The card is designed for a single-use, with a series of capture and purification membranes, to facilitate the purification of anion exchange–quality plasmid DNA starting directly from culture.

The card is designed for a single-use, with a series of capture and purification membranes, to facilitate the purification of anion exchange–quality plasmid DNA starting directly from culture. The reagent tray is pre-packed and foil-sealed with all necessary buffers. Disposable cell liners and lids are also provided for adding bacterial culture into the instrument. No additional reagents or plasticware is needed for the plasmid purification.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Bacterial Culture
Format Proprietary Card
Capacity 1 Prep/Card/Reagent Tray
Content And Storage • BenchPro™ 2100 Plasmid Purification Cards, 1 pack of 4
•Cell Liners, 1 pack of 4
• Cell Liner Lids, 1 pack of 4
• Elution Tubes (2 mL), 1 pack of 4
• Reagent Trays (prefilled and foil-sealed with reagents), 4 trays
For Use With (Equipment) BenchPro™ 2100 Plasmid Purification System
Includes BenchPro™2100 Plasmid Purification Cards 1 pack of 4; Cell Liners 1 pack of 4; Cell Liner Lids 1 pack of 4; Elution Tubes (2 mL) 1 pack of 4; Reagent Trays (prefilled and foil-sealed with reagents) 4 trays
Product Line BenchPro
Quantity 4 Sets/Box
Run Time 90 min.
Sample Volume 100 to 125 mL bacterial culture
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.