missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BioPrime™ Total Genomic Labeling System

Product Code. 10343803
Change view
Click to view available options
Quantity:
10 Reactions
30 Reactions
Unit Size:
30 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10343803 30 Reactions Each
10109972 10 Reactions 30 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10343803 Supplier Invitrogen™ Supplier No. 18097011
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Complete genomic DNA labeling kits designed for use in array comparative genomic hybridization applications (aCGH) that allows for better call rates with precious samples

These complete, all-inclusive kits are optimized to work across the widest range of sample input material with no need for pre-amplification. Highly concentrated exo-Klenow fragment of DNA polymerase I in a component limited reaction allow for consistent, robust DNA yields of ∽8μg with as little as 50ng of genomic DNA input. An optimized dye labeled nucleotide mix with new dNTP linkers, novel, application-specific Alexa Fluor 3 and 5 dyes, and improved buffer chemistry reduce labeling variation and increase signal to noise on arrays. Excitation and emission spectra of Alexa Fluor 3 and 5 dyes are suited for conventional two color scanners with no need to change settings. Complete kits are batch tested to assure quality in performance from lot to lot.
Contents and Storage:
The simplified kits contains Alexa Fluor 3 and Alexa Fluor 5 2x reaction mix, Exo-Klenow fragment (40U/μL), 5mM EDTA, TE Buffer, and control DNA (Salmon Sperm). The BioPrime Total Genomic Labeling System also contains PurelinkTM Genomic DNA purification columns and purification buffers (the BioPrime Total Genomic Labeling Module does not). New master mix formulation streamlines reaction set-ups, increasing consistency.

  • Higher signal to noise with Alexa Fluor™ 3 and 5 dyes
  • Less channel bias due to novel reaction formulation
  • Widest range of input material (50ng - 3μg)
  • Simplified workflow with master mix formulation

Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Non-Coding RNA Research

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Label or Dye Alexa Fluor 3, Alexa Fluor 5
Final Product Type Probes (Labeled DNA)
Labeling Target Genomic DNA
Includes Label or Dye Yes
Labeling Method Direct Labeling
Product Line BioPrime
Product Type Total Genomic Labeling System
Quantity 30 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.