missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calcium Phosphate Transfection Kit

Product Code. 10124462
Click to view available options
Quantity:
75 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10124462

Brand: Invitrogen™ K278001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

2 x 12mL Tissue Culture Sterile Water, 2 x 12mL 2X HEPES Buffered Saline (HBS), 3 x 1mL 2M CaCl2, 20μg pcDNA™3.1/His/lacZ

Provides high-quality reagents to enable the introduction of DNA into eukaryotic cells via calcium phosphate co-precipitation

  • Based on forming a calcium phosphate-DNA precipitate
  • Facilitates the binding of the DNA to the cell surface. DNA then enters the cell by endocytosis
  • First developed by Graham and van der Ebb and was later modified by Wigle
  • Used to transfect a wide variety of cell types for transient expression or for producing stable transformants
  • DNA is mixed directly with a concentrated solution of CaCl2, which is then added dropwise to a phosphate buffer to form a fine precipitate
  • Aeration of the phosphate buffer while adding the DNA-CaCl2 solution helps to ensure that the precipitate that forms is as fine as possible, which is important because clumped DNA will not adhere to or enter the cell as efficiently
  • Product size: 75 reactions

Cell Culture, Plasmid Transfection, Transfection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 × 12 ml Tissue Culture Sterile Water
2 × 12 ml 2X HEPES Buffered Saline (HBS)
3 × 1 ml 2 M CaCl2
20 μg pcDNA™3.1/His/lacZ

Store all components at -5 to -30°C.
Cell Type Established Cell Lines
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA
Transfection Technique Chemical Transfection
For Use With (Application) Transfection
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Type Transfection Kit
Quantity 75 Reactions
Serum Compatible No

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.