Learn More
Invitrogen™ Champion™ pET151 Directional TOPO™ Expression Kit with BL21 Star™ (DE3) One Shot™ Chemically Competent E. coli
Product Code. 10423822
Change view
Click to view available options
Quantity:
20 reactions
Unit Size:
Each
Product Code. Quantity Unit Size
10423822 20 reactions Each
1 options
Product Code. 10423822 Supplier Invitrogen™ Supplier No. K15101
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
The Directional TOPO Expression box contains 200ng of linearized, topoisomerase I-activated Champion pET vector; sterile water; dNTPs; 10X PCR Buffer; salt solution; control template and primers; primers for sequencing or PCR screening; and an expression control. The BL21 Star(DE3) box contains 21 x 50μL aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid.
Protein of interest can reach levels greater than 50 percent of total cellular protein during expression
  • Based on T7 expression vectors originally developed by Studier and colleagues, high-level expression is achieved because the T7 RNA polymerase is more processive than native E. coliRNA polymerase and is dedicated to transcription of gene of interest
  • Protein production is further enhanced in system by expression strain BL21 StarE. coli, which significantly improves stability of mRNA transcripts and increases protein expression up to ten-fold
  • Simplified, efficient Directional TOPO cloning allows to quickly enter Champion pET Expression vector
  • Feature linearized, topoisomerase I-activated Champion pET expression vectors for five-minute directional cloning
  • Directional TOPO Cloning technology facilitates gene expression because proofreading enzyme is used for PCR, resulting in fewer errors in cloned genes
  • Greater than 90% of clones are in correct orientation for gene expression, reducing time spent on colony screening
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Constitutive or Inducible System Inducible
Inducing Agent IPTG
Promoter T7, lacO
Product Type TOPO Expression Kit
Selection Agent (Eukaryotic) None
Content And Storage Each Champion™ pET Directional TOPO™ Expression Kit is provided as a complete expression system. The Directional TOPO™ Expression box contains 200 ng of linearized, topoisomerase I-activated Champion™ pET vector; sterile water; dNTPs; 10X PCR Buffer; salt solution; control template and primers; primers for sequencing or PCR screening; and an expression control. Store at -20°C. The One Shot™ TOP10 box contains twenty-one 50-μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. The One Shot™ BL21 Star™(DE3) box contains twenty-one 50-μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. Kits with Lumio™ Technology include 20 μl of the Lumio™ Detection Reagent. Store at -20°C. Guaranteed stable for 6 months when properly stored.
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Bacterial or Yeast Strain BL21 Star™(DE3)
Cleavage TEV Protease Recognition Site
Expression System E. coli
Expression Mechanism Cell-Based Expression
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method Directional TOPO
Quantity 20 reactions
Vector pET
Product Line One Shot