Learn More
Invitrogen™ Champion™ pET200 Directional TOPO™ Expression Kit with BL21 Star™ (DE3) One Shot™ Chemically Competent E. coli
Product Code. 10634223
Change view
Click to view available options
Quantity:
20 reactions
Unit Size:
Each
Product Code. Quantity Unit Size
10634223 20 reactions Each
1 options
Product Code. 10634223 Supplier Invitrogen™ Supplier No. K20001
  • ONLINE EXCLUSIVE PRICE
    1364.00 €
    1118.48 € / 20 reactions
    Save 245.52 €
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
200ng of linearized, topoisomerase I-activated Champion pET vector; sterile water; dNTPs; 10X PCR Buffer; salt solution; control template and primers; primers for sequencing or PCR screening; and an expression control. The TOP10 box contains 21 x 50μL aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. The BL21 Star(DE3) box contains 21 x 50μL aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid.
Protein of interest can reach levels greater than 50 percent of total cellular protein in E. coli during expression
  • Based on T7 expression vectors originally developed by Studier and colleagues, high-level expression is achieved because the T7 RNA polymerase is more processive than native E. coliRNA polymerase and is dedicated to transcription of gene of interest
  • Protein production is further enhanced in system by expression strain BL21 StarE. coli, which significantly improves stability of mRNA transcripts and increases protein expression up to ten-fold
  • Simplified, efficient Directional TOPO cloning allows to quickly enter a ChampionpET Expression vector
  • These kits feature linearized, topoisomerase I-activated ChampionpET expression vectors for five-minute directional cloning to facilitate gene expression with proofreading enzyme used for PCR, resulting in fewer errors in cloned genes
  • Greater than 90% of clones are in correct orientation for gene expression, reducing the time spent on colony screening
Bacterial ExpressionProtein ExpressionProteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Constitutive or Inducible System Inducible
Inducing Agent IPTG
Promoter T7, lacO
Product Type TOPO Expression Kit
Selection Agent (Eukaryotic) None
Content And Storage Each Champion™ pET Directional TOPO™ Expression Kit is provided as a complete expression system. The Directional TOPO™ Expression box contains 200 ng of linearized, topoisomerase I-activated Champion™ pET vector; sterile water; dNTPs; 10X PCR Buffer; salt solution; control template and primers; primers for sequencing or PCR screening; and an expression control. Store at -20°C. The One Shot™ TOP10 box contains twenty-one 50-μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. The One Shot™ BL21 Star™(DE3) box contains twenty-one 50-μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. Kits with Lumio™ Technology include 20 μl of the Lumio™ Detection Reagent. Store at -20°C. Guaranteed stable for 6 months when properly stored.
Antibiotic Resistance Bacterial Kanamycin (KanR)
Bacterial or Yeast Strain BL21 Star™(DE3)
Cleavage EK (Enterokinase) Recognition Site
Expression System E. coli
Expression Mechanism Cell-Based Expression
Protein Tag His Tag (6x), Xpress Epitope Tag
Cloning Method Directional TOPO
Quantity 20 reactions
Vector pET
Product Line One Shot