missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ Forensic DNA Purification Kit

Product Code. 10493282
Change view
Click to view available options
No. of Reactions:
100 Preps
960 Preps
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10493282 100 Preps Each
10614123 960 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10493282 Supplier Invitrogen™ Supplier No. CS11200
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

ChargeSwitch Lysis Buffer (L13)
ChargeSwitch Magnetic Beads
Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
ChargeSwitch Purification Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

ChargeSwitch™ Forensic DNA Purification Kit uses magnetic bead technology to allow for rapid and efficient purification of genomic DNA from a variety of forensic samples, including poor or environmentally degraded samples, and is fully validated for Single Tandem Repeat (STR) analysis.

ChargeSwitch™ Forensic DNA Purification Kit uses magnetic bead technology to allow for rapid and efficient purification of genomic DNA from a variety of forensic samples, including poor or environmentally degraded samples, and is fully validated for Single Tandem Repeat (STR) analysis.

• Optimized for higher-purity DNA extraction
• Validated for STR profiling on diverse forensic samples-set
• Compatible with high-throughput protocols

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

All components of the ChargeSwitch™ Forensic DNA Purification Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly.

Components:
• ChargeSwitch™ Lysis Buffer (L13)
• ChargeSwitch™ Magnetic Beads
• Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
• ChargeSwitch™ Purification Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Forensic Sample
For Use With (Application) Forensic
High-throughput Compatibility High-throughput Compatible
No. of Reactions 100 Preps
Product Line ChargeSwitch
Product Type Forensic DNA Purification Kit
Quantity 100 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Test Time 15 min.
Show More Show Less
compliance-icons
Product Identifier
  • ChargeSwitch Forensic DNA Purification Kit
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Proteinase, tritirachium album serine

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.