missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™-Pro PCR Clean-Up Kit

Product Code. 10542894
Change view
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
10542894 50 Preps Each
10357943 10 Preps Each
10430164 250 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10542894 Supplier Invitrogen™ Supplier No. CS32050
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.

• 100% ethanol-free, guanidinium-free, organic solvent-free protocols
• Improved downstream performance
• Complete kit for fast, hassle-free setup
• 20% fewer handling steps than most PCR clean-up protocols
• Purification of up to 500 μL of PCR product in one column

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

The ChargeSwitch™-Pro PCR Clean-up Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.

Store at room temperature. Guaranteed stable for 6 months when properly stored.

Format Kit
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line ChargeSwitch
Product Type PCR Clean-Up Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Target DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application) Next-Generation Sequencing (NGS), Cloning, Standard PCR
Isolation Technology ChargeSwitch™ Chemistry
No. of Reactions 50 Preps
Sample Type PCR Products
Show More Show Less
compliance-icons
Product Identifier
  • CHARGESWITCH PRO PCR
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Hydrogen chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.