missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CIAP (Calf Intestinal Alkaline Phosphatase), 20 U/μL

Product Code. 10747864
Change view
Click to view available options
Quantity:
1000 U
Unit Size:
1ku
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10747864 1000 U 1ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10747864 Supplier Invitrogen™ Supplier No. 18009019
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phosphomonoesterase that removes 3' and 5' phosphates from DNA and RNA

  • Concentration: 20 units/μL
  • Source: Purified from calf intestinal mucosa
  • Performance and Quality Testing: Endodeoxyribonuclease, 3' exodeoxyribonuclease, and ribonuclease assays; dephosphorylation efficiency measured in a transformation assay
  • Unit Definition: One unit hydrolyzes 1μmol of 4-nitrophenyl phosphate in 1 min. at 37°C
  • Unit Reaction Conditions: 1M diethanolamine buffer, 10mM 4-nitrophenyl phosphate, 0.25mM MgCl2 (pH 9.8) in 900μL for 10 min. at 37°C

Dephosphorylation of 5'-phosphorylated termini of vector DNA to prevent self-ligation; Dephosphorylation of 5' termini of nucleic acids prior to forward reaction with kinase; Cloning; Restriction Enzyme Cloning

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 U/μL
Content And Storage Calf Intestinal Alkaline Phosphatase (20 units/μL) is supplied with a vial of 10X dephosphorylation buffer [500 mM Tris-HCl (pH 8.5), 1 mM EDTA], vial of dilution buffer [50% glycerol, 25 mM Tris-HCl (pH 7.6), 1 mM MgCl2 , and 0.1 mM ZnCl2 ]. Store Calf Intestinal Alkaline Phosphatase at -20°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme CIP
Quantity 1000 U
Product Type Alkaline Phosphatase

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.