Learn More
Invitrogen™ Daughter E-Base™ device
Description
Includes
One Daughter E-Base™ Device
Each device features a small foot-print for space saving, and a built-in power supply.
- The Daughter E-Base device does not connect with a power cord and cannot be used without a Mother E-Base device
- Each E-Base device has a power/program button and a timer button on the lower right side for controlling the electrophoresis run
- The lower left side of each device contains a lighted LED and a digital timer display (00-99) that indicates where your gel is in the running process
- Two programs are pre-set in the E-Base devices, one each for E-Gel and E-PAGE gels
- The E-PAGE 96 gel is not compatible with older models of the devices (E-Gel 96 mother and daughter bases)
- The older models cannot be used in combination with the Mother and Daughter E-Base devices
- E-Gel 48 Medium Throughput DNA Electrophoresis System E-Gel 48 gels are ready-to-use, precast agarose gels designed for medium throughput analysis of PCR products and DNA fragments
- All E-Gel gels include a gel matrix and electrodes packaged inside a disposable, UV-transparent cassette
- E-Gel 48 gels are incorporated with ethidium bromide, and contain 2 rows of wells, with 24 sample wells and 2 marker wells per row
- Samples have a run length of 3.2 cm. E-Gel 48 gels can be loaded using multichannel pipettors or automated liquid handling systems
- DNA bands resolve clearly in just 20 minutes or less
- For precise and effortless tracking of gels, each cassette is individually labeled with an EAN39 bar code
Two types of bases are offered:
- 1. The Mother E-Base™ device comes with a power cord that can be connected directly to an electrical outlet and is used for electrophoresis of one gel
- 2. The Daughter E-Base device connects to the Mother E-Base and to other Daughter E-Base devices for the simultaneous electrophoresis of two or more gels
- Ready-to-use, precast E-Gel 96 gels are designed for fast, high throughput DNA electrophoresis
- Like all E-Gel gels, E-Gel 96 gels include a gel matrix and electrodes packaged inside a disposable, UV-transparent cassette
- Each gel contains 96 sample wells and 8 marker wells arranged in a patented staggered format that provides samples with a 1.6 cm run length
- The gels are compatible with 8-, 12-, and 96-tip automated loading robots to provide optimum performance and resolution
- With just 12-minute run times, up to 20,000 samples can be run in a single day, saving weeks of screening time
- As with E-Gel 48 gels, each cassette is individually labeled with an EAN39 bar code that is recognized by most commercially available robotic bar code readers
- E-Gel 96 gels are offered with a choice of ethidium bromide or SYBR™ Safe DNA Gel Stain incorporated in them
- The E-PAGE High-Throughput (HTP) Protein Electrophoresis System is the only system available for fast, bufferless, high-throughput protein analysis
- It consists of E-PAGE 96 gels, E-Base™ integrated devices, the E-Holder™ platform, and the free E-Editor™ 2.0 software
- E-PAGE 96 gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable, UV-transparent cassette
- Each E-PAGE 96 gel contains 96 sample lanes and 8 marker lanes in a patented staggered well format with a 1.6 cm run length
- The well openings of the E-PAGE 96 cassette are compatible with the standard 96-well plate format and can be conveniently loaded with a multichannel pipettor or 8-, 12-, or 96-tip liquid-handling robotic devices
- E-PAGE 96 cassettes are conveniently opened with a butterfly opener to remove the gel for staining or blotting. In addition, each E-PAGE 96 cassette is labeled with an individual barcode
- The E-Editor 2.0 software is designed for fast and convenient analysis of results from E-Gel 96 and E-PAGE 96 gels and blots. Using the digital image of the gel, the software reconfigures the staggered lanes into one regrouped and aligned image for easy comparison, documentation, and reference with the original plate coordinates, allowing quick identification of important results
- The E-Holder platform is compatible in size with standard automated systems for convenient loading of samples in a hands-free robot environment when the use of the E-Base devices is not practical or while in queue for its availability
1D Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Content And Storage | One Daughter E-Base™ Device |
| For Use With (Equipment) | E-Base™ |
| Product Line | Novex |
| Type | Daughter E-Base Device |
| Quantity | 1 unit |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.