missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDM (n-dodecyl β-D-maltoside) (10%)

Product Code. 10004092
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10004092 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10004092 Supplier Invitrogen™ Supplier No. BN2005
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Detergent solution used to prepare native protein samples for use with the NativePAGE™ Gel System

  • Ready-to-use detergent solution improves solubility of hydrophobic and membrane proteins during sample preparation
  • The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis showing increased resolution and reduced streaking

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 ml of solution. Store at +4°C.
Form Liquid
Quantity 1 mL
Reagent Type DDM
Shipping Condition Wet Ice
Product Type DDM

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.