missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD8 Positive Isolation Kit

Product Code. 10527213
Click to view available options
Quantity:
5 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10527213

Brand: Invitrogen™ 11333D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

5mL Dynabeads CD8 coated with anti-CD8 monoclonal antibody and 2mL DETACHaBEAD™ CD8.

The Dynabeads™ CD8 Positive Isolation Kit supplies superparamagnetic beads that enable gentle isolation of pure CD8+ T cells from whole blood, bone marrow, buffy coat, mononuclear cells (MNC), or tissue digests.

The Dynabeads™ CD8 Positive Isolation Kit supplies superparamagnetic beads that enable gentle isolation of pure CD8+ T cells from whole blood, bone marrow, buffy coat, mononuclear cells (MNC), or tissue digests. The cells are released from the beads using the DETACHaBEAD™ reagent to obtain bead- and antibody-free cells. Advantages of the Dynabeads™ CD8 Positive Isolation Kit:

• Gentle isolation of CD8+ T cells from any sample—no columns required
• Bead- and antibody-free positive isolation of CD8+ cells for any downstream assay

About Dynabeads™ CD8 Positive Isolation Kit
The Dynabeads™ CD8 Positive Isolation Kit contains uniform, superparamagnetic beads (4.5 μm diameter) covalently coated with a monoclonal antibody specific for the CD8 membrane antigen predominantly expressed on the cytotoxic subset of human T cells. The kit also includes a DETACHaBEAD™ reagent that releases the Dynabeads™ from the purified CD8+ cells. This gently magnetic isolation process gives highly viable bead- and antibody-free cells to be used in any downstream assay.

Magnetic bead–based separation offers easy handling
Dynabeads™ are mixed with the sample in a tube where they bind to the target cells during a short incubation, after which the bead-bound cells are separated from unbound cells using a magnet. The positively isolated cells are then washed, and DETACHaBEAD™ reagent is added to gently release the cells from the beads. This magnetic bead-based positive isolation method gives both bead- and antibody-free cells. The isolated CD8+ cells are phenotypically unaltered and suitable for any downstream application. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity and viability of the isolated CD8+ cells. For depletion or isolation of untouched CD8+ cells, select from our range of Human CD8+ T Cell Isolation Products.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (CD8)
Content And Storage This Product Contains: 5 mL Dynabeads™ CD8 coated with anti-CD8 monoclonal antibody and 2 mL DETACHaBEAD™ CD8.

Storage Conditions: 2°C to 8°C. Do not freeze.
Product Type Positive Cell Isolation Kit
Isolation Technology Positive Isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DETACHaBEAD, DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Sample Type PBMC, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species Human
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.