Learn More
Invitrogen™ DMRIE-C Transfection Reagent
Description
Includes
Contains one tube (1mL) DMRIE-C Transfection Reagent.
DMRIE-C Transfection Reagent is particularly effective for transfecting suspension cells (e.g., Jurkat) and other lymphoid-derived cell lines. DMRIE-C can also be used for in vivo delivery of DNA. DMRIE-C is a 1:1 (M/M) liposome formulation of the cationic lipid DMRIE (1,2-dimyristyloxy-propyl-3-dimethyl-hydroxy ethyl ammonium bromide) and cholesterol in membrane-filtered water.
DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, RNA Extraction, Total RNA Isolation, Total RNA from Liquid Samples (e.g. Serum, Virus), Viral RNA/DNA Purification, gDNA from Virus Samples
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Content And Storage | Contains one tube (1 ml) DMRIE-C Transfection Reagent. Store at 4°C. Do not freeze. |
| Cell Type | Established Cell Lines |
| Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
| Sample Type | Plasmid DNA |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | Transfection |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Product Type | Transfection Reagent |
| Quantity | 1 mL |
| Serum Compatible | No |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.