missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Plant DNAzol™ Reagent

Product Code. 11598836
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11598836 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11598836 Supplier Invitrogen™ Supplier No. 10978021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Plant DNAzol™ Reagent is a ready-to-use organic reagent formulated for the isolation of high quality genomic DNA from a variety of plant samples.

Plant DNAzol™ Reagent is a ready-to-use organic reagent formulated for the isolation of high quality genomic DNA from a variety of plant samples.

  • Permits use on broad spectrum plant material
  • Rapid isolation of genomic DNA

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 100 mL; room temperature

Isolation Technology Organic Extraction
Sample Type Plants
Elution Volume 70 μL
Final Product Type Genomic DNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 100 mL
Shipping Condition Room Temperature
Test Time 60 min.
Yield ≤300 μg
Starting Material Amount 0.1 g
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.