missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CELLection™ Pan Mouse IgG Kit

Product Code. 10252233
Click to view available options
Quantity:
5 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10252233

Brand: Invitrogen™ 11531D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

CELLection Dynabeads coated with a human anti-mouse IgG monoclonal, and DNase Releasing Buffer

Use this kit with any mouse IgG for the positive isolation of a cell population from all species except mouse.

Use this kit with any mouse IgG for the positive isolation of a cell population from all species except mouse. Antibodies are attached to the surface of the CELLection™ Dynabeads™ via a DNA linker; this linker provides a cleavable site to release and remove the beads from the cells after isolation. Bead-free cells are viable and can be used in any downstream application.

This Kit Contains
CELLection™ Dynabeads™ coated with a human anti-mouse IgG monoclonal, and DNase Releasing Buffer

Applications
• Positively isolate and detach any cell from any species (except mouse) using your own mouse IgG
• Compatible with flow cytometry, cell culture and all cell based assays

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Antibody Fragment Whole Antibody
Cell Type All cells from all species, except mouse
Content And Storage Store in refrigerator (2–8°C).
Product Type Antibody Coated Bead
Dye Type CELLection™ Pan
Host Species Mouse
Isolation Technology Positive Isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line CELLection, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Reactivity All species, except mouse
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species All species, except mouse
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.