missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ M-270 Amine

Product Code. 10151003
Change view
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10151003 2 mL Each
10161003 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10151003 Supplier Invitrogen™ Supplier No. 14307D
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These are hydrophilic Dynabeads™ (2.8 μm) with amine groups. The surface amino groups allow direct covalent binding of carbohydrates, glycoproteins and glycolipids through reductive amination of aldehydes.

The surface amino groups allow direct covalent binding of carbohydrates, glycoproteins and glycolipids through reductive amination of aldehydes. For use with any sample type.

  • Rapid binding (less than 1 hour) at neutral to high pH and room temperature
  • No further surface activation required
  • Excellent reactivity with a range of cross-linking reagents, allowing for easy introduction of further alternative surface chemistries
  • Ideal for isolation of small components such as proteins and peptides
  • Allows C-terminal coupling of proteins
  • Well suited for isolation of proteins or other targets with affinity for specific carbohydrate moieties

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Diameter (Metric) 2.8 μm
Elution Volume Min 5 μL
High-throughput Compatibility High-throughput Compatible
Shipping Condition Room Temperature
Sample Type Any Sample Type
Surface Functionality Amine
Product Line Dynabeads
Quantity 2 mL
Type Bead with Amine Group
Format Beads in suspension
Purity or Quality Grade Research Grade
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.