missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome-Human CD81 Flow Detection Reagent (from cell culture)

Product Code. 15490544
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
This item is not returnable. View return policy

Product Code. 15490544

Brand: Invitrogen™ 10622D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell culture media must first be enriched for exosomes using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation.

Cell culture media must first be enriched for exosomes using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation. Exosome-Human CD81 Flow Detection Reagent (from cell culture) allows easy visualization of exosomes by flow cytometry using this magnetic separation technology. Free exosomes alone are too small to be detected by most flow cytometry instruments. The 2.7 μm Dynabeads magnetic beads allow for a clear and defined flow analysis.

  • Minimal hands-on time for specific exosome capture and analysis
  • Clear and defined flow analysis in less than 1 hour hands-on time
  • Study expression of specific exosome subset markers
  • Protocol is easily scalable
  • Recent studies suggest that exosome/ECV population consists of several subsets, possibly with different functions
  • Using immuno-affinity based Dynabeads magnetic separation technology, these subsets can be selectively captured and studied using flow cytometry and/or electron microscopy
  • Dynabeads technology avoids loss of material due to handling and centrifugation typically experienced with other isolation and analysis methods

Mix thoroughly to ensure that magnetic beads do not settle in the sample tube. A mixer which provides both tilting and rotation for optimal mixing is recommended.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Product Type Flow Detection Reagent
Product Line Dynabeads
Quantity 2 mL
Sample Type Cell Cultures
Diameter (Metric) 2.7 μm

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.