Learn More
Invitrogen™ Exosome-Human EpCAM Flow Detection Reagent (from cell culture)
Description
Cell culture media must first be enriched for exosomes using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation. Exosome-Human EpCAM Flow Detection Reagent (from cell culture) allows easy visualization of exosomes by flow cytometry using this magnetic separation technology. Free exosomes alone are too small to be detected by most flow cytometry instruments. The 2.7 μm Dynabeads magnetic beads allow for a clear and defined flow analysis.
- Minimal hands-on time for specific exosome capture and analysis
- Clear and defined flow analysis in less than 1 hour hands-on time
- Study expression of specific exosome subset markers
- Protocol is easily scalable
- Recent studies suggest that exosome/ECV population consists of several subsets, possibly with different functions
- Using immuno-affinity based Dynabeads magnetic separation technology, these subsets can be selectively captured and studied using flow cytometry and/or electron microscopy
- Dynabeads technology avoids loss of material due to handling and centrifugation typically experienced with other isolation and analysis methods
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C). |
| Product Type | Flow Detection Reagent |
| Product Line | Dynabeads |
| Quantity | 2 mL |
| Sample Type | Cell Cultures |
| Diameter (Metric) | 2.7 μm |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.