missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome-Human CD9 Isolation Reagent (from cell culture)

Product Code. 15450544
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450544 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450544 Supplier Invitrogen™ Supplier No. 10614D

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Obtain a highly pure exosome subset with easy protocol scalability with Exosome-Human CD9 Isolation Reagent (from cell culture).

Obtain a highly pure exosome subset with easy protocol scalability with Exosome-Human CD9 Isolation Reagent (from cell culture). Following isolation, the specific sub-population of exosomes, ECVs can be further studied using methods such as Western blotting, qPCR, or next generation sequencing.

  • Minimal hands-on time for specific exosome isolation
  • Compatible with most downstream analysis
  • Recent studies suggest that exosome/ECV population consists of several subsets, possibly with different functions
  • Using immuno-affinity based Dynabeads magnetic separation technology, these subsets can be selectively isolated and studied
  • Cell culture media must first be enriched for exosomes
  • Can be done quickly and efficiently using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation
  • Following enrichment, specific exosomes are isolated by binding to surface of magnetic beads
  • In addition to their high-quality sensitivity, reproducibility, and stability, Dynabeads allows you to see samples during handling due to color of magnetic beads
  • When a sample tube is placed in the magnet, bead-bound exosomes are pulled to side of tube, allowing for easy separation and buffer changes
  • Sample and bead volume can be easily scaled according to your sample size or downstream applications

Mix thoroughly to ensure that magnetic beads do not settle in the sample tube. A mixer which provides both tilting and rotation for optimal mixing is recommended.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Product Type Isolation Reagent
Product Line Dynabeads
Quantity 2 mL
Sample Type Cell Cultures

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.