Learn More
Invitrogen™ Dynabeads™ CD31 Endothelial Cell
Description
Includes
5mL Dynabeads coated with anti-CD31 monoclonal antibody
Isolate endothelial cells directly from tissue digests. Dynabeads™ CD31 are ideal for positively isolating endothelial cells for downstream molecular analysis.
• Positively isolate endothelial cells directly from tissue digests for molecular applications
• Establish cultures for endothelial cells
• No columns required
Starting sample:
Tissue digests.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Type | Endothelial Cells |
| Content And Storage | This Product Contains: 5 mL Dynabeads™ coated with anti-CD31 monoclonal antibody. Store at 2°C to 8°C. |
| Product Type | Antibody Coated Bead |
| Isolation Technology | Positive Isolation |
| No. of Cells | Processes ∼2 x 109 cells total |
| Output Viability | >95% |
| Product Line | DYNAL, Dynabeads |
| Purity or Quality Grade | Research Grade |
| Quantity | 5 mL |
| Sample Type | Tissue Digests |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.