missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ FlowComp™ Flexi Kit

Product Code. 10073193
Click to view available options
Quantity:
3 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10073193

Brand: Invitrogen™ 11061D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

3mL FlowComp; Dynabeads and 40mL FlowComp; Release Buffer and a complete DSB-X; Biotin Protein Labeling Kit

Allows isolatation of practically any cell type from any species or starting sample using selected antibody.

Allows isolatation of practically any cell type from any species or starting sample using selected antibody. Immediately after positive isolation, the beads are released and removed from your cell sample. In this way, you avoid exposing your cells to potentially cytotoxic or immunogenic foreign substances like iron oxides or dextrans, a potential problem when working with biodegradable particles. In addition, your cells will not be exposed to the stress of being passed through a column. The kit supplies everything you need for DSB-X; biotinylation of antibodies and subsequent isolation of cell type of interest. This technology has been successfully used to isolate number of cell subsets including human and mouse T cells, B cells, NK cells, monocytes, and endothelial cells.

  • Combine with your antibody of choice to isolate any cell type
  • No risk of results being affected by isolation method
  • Gentle tube-based method—no columns needed
  • High purity, recovery and viability
  • Bead-free cells—compatible with flow cytometry analysis or for direct use in any cell-based assay

Please note that the choice of antibody clone is of key importance for successful isolation (see package insert for technical recommendations).

TRUSTED_SUSTAINABILITY

Specifications

Cell Type All cells from all species
Content And Storage This Kit Contains: 3 mL FlowComp™ Dynabeads™ and 40 mL FlowComp™ Release Buffer and a complete DSB-X™ Biotin Protein Labeling Kit

Store at: 2°C to 8°C
Product Type Cell Isolation Kit
Isolation Technology Positive Isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads, FlowComp
Purity or Quality Grade Research Grade
Quantity 3 mL
Reactivity All species
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species All species
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.