missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynabeads™ FlowComp™ Human CD14 Kit
Click to view available options
Quantity:
2 x 3 mL
Unit Size:
Each
Description
Includes
2 x 3mL FlowComp; Dynabeads, 1mL FlowComp; Human CD14 Antibody and 2 x 20mL FlowComp; Release Buffer
The cells can be released from beads using the FlowComp Release Buffer.
- Rapid isolation of CD14+ monocytes from any sample - no columns required
- Pure and bead-free CD14+ monocytes for downstream application
- Not high throughput compatible
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Type | Monocytes |
| Clonality | Monoclonal |
| Content And Storage | The Kit Contains: • 2 x 3 mL FlowComp™ Dynabeads™ • 1 mL FlowComp™ Human CD14 Antibody • 2 x 20 mL FlowComp™ Release Buffer. Store at: 2°C to 8°C. |
| Product Type | Superparamagnetic Beads |
| Final Product Type | Cells |
| Format | Beads in solution |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Isolation Technology | Positive isolation - cells are left bead-free after isolation |
| Ligand Type | Antibody |
| No. of Cells | 2 x 109 PBMC, 80 mL whole blood, or 160 mL buffy coat |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction