missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ FlowComp™ Human CD4 Kit

Product Code. 10144833
Click to view available options
Quantity:
3 mL
Unit Size:
3mL
This item is not returnable. View return policy

Product Code. 10144833

Brand: Invitrogen™ 11361D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

1mL Antibody, 3mL FlowComp; Dynabeads and 40mL FlowComp; Release Buffer

This gentle and easy-to-use kit allows you to isolate flow-compatible human CD4+ T-cells from PBMC or directly from whole blood.

This gentle and easy-to-use kit allows you to isolate flow-compatible human CD4+ T-cells from PBMC or directly from whole blood. Immediately after positive isolation, the beads are released and removed from your cell sample. In this way, you avoid exposing your cells to potentially cytotoxic or immunogenic foreign substances like iron oxides or dextrans, a potential problem when working with biodegradable particles. In addition, your cells will not be exposed to the stress of being passed through a column.
You can expect a purity level of >95% and up to 75% recovery of human CD4+ cells.

Advantages
• No risk of your results being affected by the isolation method
• Gentle tube-based method—no columns needed
• High purity and recovery
• Bead-free cells—compatible with flow cytometry analysis
• Viable, bead-free CD4+ T-cells for direct use in any cell-based assay

The isolated cells have the presumed profile of memory and naive CD4+ T cells, and show a high proliferating and functional capacity. This kit avoids contamination of your isolated CD4+ cells with CD14+ monocytes. You will save time and money, and get increased confidence in your results.

Starting sample
MNC or whole blood

Capacity:
The kit will process 2 × 109 mononuclear cells (MNC) or 80 mL whole blood.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (CD4)
Content And Storage The Kit Contains:
• 1 mL Antibody
• 3 mL FlowComp™ Dynabeads™
• 40 mL FlowComp™ Release Buffer.

Store at: 2°C to 8°C.
Product Type Superparamagnetic Beads
Isolation Technology Positive Isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads, FlowComp
Purity or Quality Grade Research Grade
Quantity 3 mL
Sample Type PBMC, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 5 x 107 PBMCs, or 2 mL whole blood per isolation
Target Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.