missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ FlowComp™ Mouse CD4 Kit

Product Code. 10574034
Click to view available options
Quantity:
3 mL
Unit Size:
3mL
This item is not returnable. View return policy

Product Code. 10574034

Brand: Invitrogen™ 11461D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

1mL Antibody Mix, 3mL FlowComp; Dynabeads and 40mL FlowComp™ Release Buffer

This gentle and easy-to-use kit allows you to isolate flow-compatible mouse CD4+ T cells. Immediately after positive isolation, the beads are released and removed from your cell sample.

This gentle and easy-to-use kit allows you to isolate flow-compatible mouse CD4+ T cells. Immediately after positive isolation, the beads are released and removed from your cell sample. In this way, you avoid exposing your cells to potentially cytotoxic or immunogenic foreign substances like iron oxides or dextrans, a potential problem when working with biodegradable particles. In addition, your cells will not be exposed to the stress of being passed through a column.

You can expect a purity level of >85% and viability of >85% of mouse CD4+ cells.

Advantages
• No risk of your results being affected by the isolation method
• Gentle tube-based method - no columns needed
• High purity, recovery and viability
• Bead-free cells - compatible with flow cytometry analysis
• Viable, bead-free CD4+ T-cells for direct use in any cell-based assay.

The isolated cells have the presumed profile of memory and naive CD4+ T cells, and show a high proliferation capacity. You will save time and money, and get increased confidence in your results.

Starting sample
Spleen or lymph node cells

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (CD4)
Content And Storage The Kit Contains:
• 1 mL Antibody Mix
• 3 mL FlowComp™ Dynabeads™
• 40 mL FlowComp™ Release Buffer.

Store at: 2°C to 8°C.
Product Type Superparamagnetic Beads
Isolation Technology Positive Isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads, FlowComp
Purity or Quality Grade Research Grade
Quantity 3 mL
Reactivity Mouse
Sample Type Lymph Node, Blood, Spleen
Shipping Condition Room Temperature
Starting Material Cell No. 5 x 107 cells per isolation
Target Species Mouse
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.