missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ FlowComp™ Mouse Pan T (CD90.2) Kit

Product Code. 10399393
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10399393

Brand: Invitrogen™ 11465D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

1mL Antibody Mix, 3mL FlowComp; Dynabeads and 40mL FlowComp; Release Buffer

This gentle and easy-to-use kit allows you to isolate flow-compatible mouse Pan T (CD90.2+) cells from splenocytes or lymph node cells.

This gentle and easy-to-use kit allows you to isolate flow-compatible mouse Pan T (CD90.2+) cells from splenocytes or lymph node cells. Immediately after positive isolation, the beads are released and removed from your cell sample. In this way, you avoid any influence of the beads to the cells. In addition, your cells will not be exposed to the stress of being passed through a column.

You can expect a purity level of 96-99% of mouse Pan T cells.

Advantages
• No risk of your results being affected by the isolation method
• Gentle tube-based method - no columns needed
• High purity, recovery and viability
• Bead-free cells - compatible with flow cytometry analysis
• Viable, bead-free Pan T-cells for direct use in any cell-based assay

Starting sample
Single cells from spleen or lymph node

Capacity
The kit will process 2 x 109 cells

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (Whole Population)
Content And Storage The Kit Contains:
• 1 mL Antibody Mix
• 3 mL FlowComp™ Dynabeads™
• 40 mL FlowComp™ Release Buffer.

Store at: 2°C to 8°C.
Product Type Cell Isolation Kit
Final Product Type Cells
Format Bead
Isolation Technology Positive Isolation
No. of Cells 2 x 109
Product Line DYNAL, Dynabeads, FlowComp
Purity or Quality Grade Research Grade, 97%
Quantity 1 kit
Reactivity Mouse
Sample Type Lymph Node, Spleen
Shipping Condition Room Temperature
Target Species Mouse
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.