missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Pan Mouse IgG

Artikelnummer. 10542795
missing translation for 'orderingAttributeHoverText'
Quantity:
2 mL
5 mL
5 x 5 mL
missing translation for 'unitSize'
2mL
Each
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 10542795

missing translation for 'mfr': Invitrogen™ 11041

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beinhaltet

5mL Dynabeads Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.

Dynabeads Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion.

Dynabeads Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion. It will bind all subclasses of mouse IgG and is Fc specific. Low amount of IgG is required for binding. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.

  • Depletion—cells can be depleted from a sample
  • Positive isolation—cells can be positively isolated from a sample for molecular applications
  • Negative isolation—cells can be negatively isolated from a PBMC sample by adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible with flow cytometry.

Note:

For cellular assays we recommend positive isolation followed by release of beads from the cells. Recommended product for bead-free cells: Dynal CELLection Pan Mouse IgG (Cat. No. 115-31D) in combination with your mouse IgG.

Starting samples:

Whole blood, PBMC, buffy coat, tissue digests.

Dynabeads Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion. It will bind all subclasses of mouse IgG and is Fc specific. Low amount of IgG is required for binding. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.

  • Depletion - cells can be depleted from a sample
  • Positive isolation - cells can be positively isolated from a sample for molecular applications
  • Negative isolation - cells can be negatively isolated from a PBMC sample by adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible with flow cytometry.

Note:

For cellular assays we recommend positive isolation followed by the release of beads from the cells. Recommended product for bead-free cells: CELLection Pan Mouse IgG (Cat. No. 115-31D) in combination with your mouse IgG.

Starting samples:

Whole blood, PBMC, buffy coat, tissue digests.

Bestellinformation

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Spezifikation

Antibody Fragment Whole Antibody
Cell Type All cells from all species, except mouse
Clonality Monoclonal
Content And Storage This Product Contains: 5 mL Dynabeads™ Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is a unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.
Store at 2°C to 8°C.
Product Type Antibody Coated Bead
Dye Type Dynabeads™ Pan
Host Species Human
Includes 5 mL Dynabeads Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.
Isolation Technology Depletion, negative isolation or positive isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Reactivity All species, except mouse
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species All species, except mouse
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt