missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Sheep-Anti Mouse IgG

Product Code. 10630514
Click to view available options
Quantity:
5 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10630514

Brand: Invitrogen™ 11031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

5mL Dynabeads Sheep anti-Mouse IgG coated with sheep polyclonal against mouse IgG1 and IgG2 (not IgG3).

Dynabeads™ Sheep anti-Mouse IgG binds monoclonal mouse IgG of your choice for specific cell isolation or depletion.

Dynabeads™ Sheep anti-Mouse IgG binds monoclonal mouse IgG of your choice for specific cell isolation or depletion. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.

• Depletion: cells can be depleted from a sample
• Positive isolation: cells can be positively isolated from a sample for molecular applications
• Negative isolation: cells can be negatively isolated from a PBMC sample by adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible with flow cytometry.

Note: For a more universal binding of your mouse IgG of any subclass, please use Dynabeads™ Pan Mouse Ig (Cat. No. 110-41D or 110-42D).

Starting samples:
Whole blood, PBMC, buffy coat., tissue digests.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Antibody Fragment Whole Antibody
Cell Type All cells from all species, except mouse
Clonality Polyclonal
Content And Storage This Product Contains: 5 mL Dynabeads™ Sheep anti-Mouse IgG coated with sheep polyclonal against mouse IgG1 and IgG2 (not IgG3).

Store at 2°C to 8°C.
Product Type Antibody Coated Bead
Dye Type Dynabeads™
Host Species Sheep
Isolation Technology Depletion, negative isolation or positive isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Reactivity All species, except mouse
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species All species, except mouse
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.