missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ 50 bp DNA Ladder

Product Code. 10122348
Change view
Click to view available options
Quantity:
100 applications
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10122348 100 applications 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10122348 Supplier Invitrogen™ Supplier No. 10488099
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated specifically for maximum performance on E-Gel EX gels as well as other E-Gel precast gels

Invitrogen E-Gel 50 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 50 bp to 2,500 bp. E-Gel 50 bp DNA Ladder consists of 17 individual chromatography-purified DNA fragments and has reference bands at 2500, 800, and 350 bp for easy orientation.

E-Gel 50 bp DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.

Highlights of E-Gel 50 bp DNA Ladder:

  • Performance—optimal for use on E-Gel agarose gels
  • Sharp, clear bands—chromatography purified fragments for consistent and reliable results and stable at room temperature up to six months
  • Ready-to-use—premixed with loading buffer
  • Convenient—provided with 1X E-Gel Sample Loading Buffer
  • Precise—an exact amount of DNA in each band

Product use
The double-stranded DNA ladder can be visualized on 2% E-Gel agarose gels. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2 x 1 mL E-Gel 50 bp DNA Ladder
• 1 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods, store at -20°C.
Concentration 0.025 μg/μL
Gel Type Agarose
Ready to Load Yes
Sample Loading Volume 1 mL
Size Range 50 to 2500 bp
Volume (Metric) 2 x 1000 μL
Gel Compatibility E-Gel™ agarose gels
Quantity 100 applications
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Type DNA Ladder
System Type E-Gel
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.