missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Go! Agarose Gels, 1%

Product Code. 12003589
Change view
Click to view available options
Quantity:
10 Gels/Pk.
20 Gels/Pk.
Unit Size:
Each
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12003589 20 Gels/Pk. Pack of 20
12383643 10 Gels/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12003589 Supplier Invitrogen™ Supplier No. G442001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For performing agarose gel electrophoresis with only a few samples to analyze

  • Precast agarose gel cassettes each have four wells and are premixed with a blue light excitable nucleic acid stain
  • Can be stored at room temperature and are ready to use without additional buffers or reagents
  • Designed specifically to work with the components of the E-Gel Go! System
  • For a quick check of end point PCRs, restriction digests, plasmid preps or other molecular biology reactions
  • Use proprietary stain formulated to provide the high detection sensitivity and can be visualized with traditional UV transilluminators
  • When using blue light the bands must be viewed through an amber filter
  • Sharp bands and repeatable, reliable results
  • Single comb well formatSample Volume; 10μL

  • For DNA fragments up to 4kbp

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Compatible Ladders E-Gel™ 1 Kb Plus Express Ladder
Content And Storage • Each pack contains 20 individually sealed 1% E-Gel™ Go! precast agarose gel cassettes

Store at room temperature.
Gel Percentage 1%
Gel Type E-Gel
Label or Dye SYBR Gold
Sample Loading Volume 10 μL
Separation Range 100 to 4000 bp
Wells 4-well
Quantity 20 Gels/Pk.
Shipping Condition Room Temperature
Well Design Single Comb
Product Type Go Agarose Gel
System Type E-Gel Go!
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.