missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ Go! Agarose Gels, 1%
Description
- Precast agarose gel cassettes each have four wells and are premixed with a blue light excitable nucleic acid stain
- Can be stored at room temperature and are ready to use without additional buffers or reagents
- Designed specifically to work with the components of the E-Gel Go! System
- For a quick check of end point PCRs, restriction digests, plasmid preps or other molecular biology reactions
- Use proprietary stain formulated to provide the high detection sensitivity and can be visualized with traditional UV transilluminators
- When using blue light the bands must be viewed through an amber filter
- Sharp bands and repeatable, reliable results
- Single comb well formatSample Volume; 10μL
- For DNA fragments up to 4kbp
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
| Content And Storage | • Each pack contains 10 individually sealed 1% E-Gel™ Go! precast agarose gel cassettes Store at room temperature. |
| Gel Percentage | 1% |
| Gel Type | E-Gel |
| Label or Dye | SYBR Gold |
| Sample Loading Volume | 10 μL |
| Separation Range | 100 to 4000 bp |
| Wells | 4-well |
| Quantity | 10 Gels/Pk. |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?