missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Go! Agarose Gels Starter Kit, 1%

Product Code. 11632169
Change view
Click to view available options
Quantity:
10 Pack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11632169 10 Pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11632169 Supplier Invitrogen™ Supplier No. G4401ST

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

1 x E-Gel Go! Base with power cable. 1 x 10-pack of 1% E-Gel Go! precast agarose gel cassettes. 1 x 100 application tube of E-Gel 1 Kb Plus DNA ladder. 4 x 1.25mL tubes of E-Gel loading buffer.

Waste fewer lanes when checking only a few samples

  • E-Gel 1 Kb Plus DNA ladder is an intuitive ladder for most applications
  • The included base is a portable, powerful device for running and visualizing E-Gel Go! precast agarose gels
  • Sleek design, minimal footprint and light weight
  • Base acts as a power supply to run precast agarose gels cassettes as well as a blue light transilluminator to view results in real time
  • Specifically designed to offer exceptional convenience when only a few samples need checking
  • Each gel cassette has four lanes that can be used for samples or reference standards
  • Cassettes are prestained with a high sensitivity, blue light excitable, nucleic acid stain
  • DNA ladder has 12 evenly spaced bands ranging from 1 Kb to 12 Kb, a quick orientation band at 1650 base pairs that forms a distinct doublet with the 2 Kb band, and seven bands of round sizes below 1 Kb
  • E-Gel 1 Kb Plus DNA ladder is specifically formulated with minimal salts
  • E-Gel Go! Gels contain single comb well format
  • Sample Volume; 10μL

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1%
Wells 4-well
Kit Contents 1 x E-Gel Go! Base with power cable. 1 x 10-pack of 1% E-Gel Go! precast agarose gel cassettes. 1 x 100 application tube of E-Gel 1 Kb Plus DNA ladder. 4 x 1.25mL tubes of E-Gel loading buffer.
Quantity 10 Pack
Storage Requirements Room Temperature
Product Type Agarose Gel Starter Pack

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.