Learn More
Thermo Scientific™ Invitrogen™ EVOS™ FL Auto 2 Imaging System
Description
- Speed: scan a 96-well plate in 3 fluorescent channels in less than 5 minutes
- Flexibility: customize the system with more than 20 user-changeable LED light cubes, dual cameras (monochrome and color), a variety of objectives ranging from 1.25x to 100x, and multiple vessel holders
- Time-lapse live-cell imaging: option for precise control of temperature, humidity, and gases for normoxic or hypoxic conditions allows a wide range of biological studies under physiological conditions with an onstage incubator
- Area view: move rapidly and seamlessly between low-magnification, single-field mode and high-magnification scan mode to easily define and capture the area of interest
- Automation: time-saving features such as autofocus, rapid stage movement, and automated routines help reduce time to complete experiments, allowing high throughput, high data quality, and improved experimental reproducibility
- Data analysis: optional advanced software package for quantitative and statistical analysis
Specifications
Specifications
| For Use With (Application) | Live Cell Imaging |
| For Use With (Equipment) | EVOS Light Cubes, EVOS Onstage Incubator |