missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome Spin Columns (MW 3000)

Product Code. 15382476
Click to view available options
Quantity:
30 Columns
Unit Size:
Pack of 30
This item is not returnable. View return policy

Product Code. 15382476

Brand: Invitrogen™ 4484449

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

3 bags (collection tubes, elution tubes, spin columns)

Fast, efficient removal of unincorporated dye from the labeled exosomes

Exosome Spin Columns (MW 3000) enable fast removal of unincorporated dye from exosome labeling reactions. Allow the user to perform buffer exchange on the exosomes or to remove any low molecular weight (MW ≤ 3000) admixtures from the exosome preparation such as salts, nucleotides, and short oligonucleotides. To use, simply rehydrate the spin column, centrifuge to remove the interstitial fluid, add the exosome sample to the top of the column, and centrifuge again to elute the exosomes (low molecular weight contaminants will be retained by the column).

  • Buffer exchange, desalting, removal of low molecular weight contaminants
  • Replaces more time-consuming purification methods (such as ultra-centrifugation)

Biomarker Discovery, Cell Analysis, Clinical, Exosomal Biomarker Analysis, Exosome Isolation, Exosomes, Organelle Isolation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 3 bags (collection tubes, elution tubes, spin columns). Store at room temperature.
Product Type Exosome Spin Column
Final Product Type Exosomes
Format Spin Column
Quantity 30 Columns

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.