Learn More
Invitrogen™ Expressway™ Mini Cell-Free Expression System
Descripción
Includes
Designed to perform 20 x 50 μL reactions or 1 x 1mL reaction. This system includes the IVPS E.coliExtract, IVPS reaction buffer, feed buffer, 19 amino acid mix, methionine, DNase/RNase-free water, RNase A, T7 Enzyme Mix, 2mL reaction tubes, and a positive expression control vector.
- Eliminates time-consuming steps of cell-based protein production including transformation, cell culture, and expression optimization
- In two hours, can produce protein suitable for array of downstream applications and functional studies
- Offers flexibility in experimental design
- Synthesize proteins from both circular plasmid and linear templates (e.g. PCR products)
- Perform high-throughput synthesis of proteins
- Express genes toxic to in vivosystems
- Provides all components need for optimal cell-free protein production
- As DNA template is transcribed, 5« end of mRNA is bound by ribosomes and undergoes translation as 3« end of template is still being transcribed
- Proprietary feed buffer formulation is provided with system for increased protein yields
Cell Free Protein Expression for Functional Analysis, Cell-Free (in vitro) Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Especificaciones
Especificaciones
| Promoter | T7 |
| Product Type | Cell-Free Expression System |
| Expressed Protein Type | Water Soluble Protein |
| Expression System | Cell-Free |
| Expression Mechanism | Cell-Free Expression |
| Quantity | 1 Kit |
| Product Line | Expressway |
Seguridad y manipulación
- Expressway Mini Cell-Free Expression System
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Tris (hydroxymethyl)aminomethane
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.