missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flp-In™ Complete System
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Description
Includes
The Flp-In Complete System includes the Core Kit (Cat. No. K601002), 1g Zeocin antibiotic, and 1g hygromycin B. All vectors are supplied supercoiled and lyophilized.
- Introduce of Flp Recombination Target (FRT) site into genome of mammalian cell line of choice
- Expression vector containing gene of interest is then integrated into genome via Flp recombinase-mediated DNA recombination at FRT site
- Major components include Flp-In. target site vector, pFRT/lacZeo, for generation of host cell line containing integrated FRT site
- Expression plasmid containing a FRT site linked to hygromycin resistance gene for Flp recombinase-mediated integration and selection of stable cell line expressing gene of interest under control of human cytomegalovirus (CMV) immediate-early enhancer/promoter
- Flp recombinase expression plasmid, pOG44, for expression of Flp recombinase under control of human CMV promoter
- Control expression plasmid containing chloramphenicol acetyl transferase (CAT) gene, which when cotransfected with pOG44 into the Flp-In. host cell line, expresses CAT
Bioproduction, Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Stable Cell Line Development, Targeted Integration
Specifications
Specifications
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Transfection |
| Promoter | CMV |
| Product Type | Flp-In Complete System |
| Selection Agent (Eukaryotic) | Zeocin™, Hygromycin |
| Content And Storage | The Flp-In™ Complete System includes the Core Kit (K601002), 1 g Zeocin™ antibiotic, and 1 g hygromycin B. All vectors are supplied supercoiled and lyophilized. Store hygromycin at +4°C. Store all other components at -20°C. Zeocin™ antibiotic should be protected from exposure to light. All reagents are guaranteed stable for 6 months when properly stored. |
| Protein Tag | Untagged |
| Cloning Method | Restriction Enzyme/MCS |
| Quantity | 1 kit |
| Vector | pFRT |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction