missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pcDNA™5/FRT/TO Vector Kit

Product Code. 11580856
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11580856 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11580856 Supplier Invitrogen™ Supplier No. V652020
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

20μg of each: pcDNAFRT/TO and pcDNAFRT/TO/CAT vectors.

5.1kb inducible expression vector features multiple cloning site with 10 unique restriction sites to facilitate cloning gene of interest

  • Control plasmid, pcDNA5/FRT/TO/CAT, is also included for use as positive control for transfection and expression in cell line of choice
  • Hybrid human cytomegalovirus (CMV)/TetO2 promoter for high-level, tetracycline-regulated expression of gene of interest in wide range of mammalian cells
  • FLP Recombination Target (FRT) site for Flp recombinase-mediated integration of vector into Flp-InT-REx host cell line
  • Hygromycin resistance gene for selection of stable cell lines

Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression, Targeted Integration

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Inducible
Delivery Type Transfection
Inducing Agent Tetracycline
Promoter CMV/TO
Product Type Flp-In/T-REx System Expression Vector
Selection Agent (Eukaryotic) Hygromycin
Content And Storage Contains 20 μg of each: pcDNA™FRT⁄TO and pcDNA™FRT⁄TO⁄CAT vectors. Store both vectors at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Protein Tag Untagged
Cloning Method Restriction Enzyme/MCS
Quantity 20 μg
Vector pcDNA
Product Line Flp-In, T-REx, pcDNA
For Use With (Application) Regulated Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.